Order Entry
Canada
Orders LinkContactUsLinkComponent
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences

Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences

D-PBS, Formula III with phenol red, Phosphate Buffer Saline, 0.15 M, 1 l, Bottle
Catalog # 100503-018
Supplier #:  11774-10
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Catalog # 100503-018
CAS Number:  
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences

D-PBS, Formula III with phenol red, Phosphate Buffer Saline, 0.15 M, 1 l, Bottle

CAS Number:  
Price
$103.60
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

All of our Dullbecco’s phosphate buffer saline solutions contain: potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose.

  • Some formulations may contain additional components
  • Store at room temperature

Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.

Specifications

Specifications

  • Concentration:
    0.15 M
  • Pack type:
    Bottle
  • Description:
    D-PBS, Formula III with phenol red, Phosphate Buffer Saline
  • Size:
    1 l

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.