Orders LinkContactUsLinkComponent
Disposable Base Mold, Electron Microscopy Sciences
Disposable Base Mold, Electron Microscopy Sciences
Disposable Base Mold, 5 mm, 30 x 24 mm
Catalog # 100502-000
Supplier #:  62352-30
Disposable Base Mold, Electron Microscopy Sciences
Catalog # 100502-000
Disposable Base Mold, Electron Microscopy Sciences
Disposable Base Mold, Electron Microscopy Sciences
Disposable Base Mold, 5 mm, 30 x 24 mm
Price
$282.79
Log in for availability
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our Disposable Base Mold is designed to fit all existing cassettes. Because our base molds are disposable you save time on cleaning and storing and they offer you less exposure to hazardous chemicals.
Specifications
  • Description:
    Disposable Base Mold
  • Height:
    5 mm
  • Size:
    30 x 24 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)