Orders LinkContactUsLinkComponent
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Filament New, Amray except 1200 series
Catalog # 100500-708
Supplier #:  81024
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Catalog # 100500-708
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Filament New, Amray except 1200 series
Price
$2,737.30
Log in for availability
Box of 10 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tungsten filments with a minimum tip radius for a coherent beam.
  • Rigid attachment of the filament wire to the post
  • Polished post ends
  • Rebuilt many times
  • Better long-term economy
The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. All of our filaments, new or rebuilt, are inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament. Filament rebuilding services include: reconditioning of the base, cleaning and/or replacement of necessary parts, installment of new standard loop or special filament tip, pre-centering, vacuum normalizing, and microscopic inspection.
Specifications
  • Description:
    Filament New
  • For use with:
    Amray except 1200 series
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)