Orders LinkContactUsLinkComponent
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
Catalog # 100499-706
Supplier #:  71156
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Catalog # 100499-706
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
Price
$425.88
Log in for availability
Dozen Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
  • Made from a special plastic that minimizes static
  • Complete with grid recording card
Documents
Specifications
  • Capacity:
    100 Grids
  • Description:
    Grid Storage Box
  • Dimensions:
    8 L×5 W×7 T mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)