Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Millonig's Phosphate Buffers, Electron Microscopy Sciences

Millonig's Phosphate Buffer, 0.2 M, Bottle, 1 l, 7.4 (0.1 M), 0...5 °C
Catalog # 100496-396
 :  11582-10
Millonig's Phosphate Buffers, Electron Microscopy Sciences
Catalog # 100496-396
CAS Number:  
Millonig's Phosphate Buffers, Electron Microscopy Sciences
Millonig's Phosphate Buffers, Electron Microscopy Sciences

Millonig's Phosphate Buffer, 0.2 M, Bottle, 1 l, 7.4 (0.1 M), 0...5 °C

CAS Number:  
Price
$98.41
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Millonig recommended to use this buffers for the fixation of very hydrated tissues with osmium tetroxide. For marine organisms, a higher concentration of sodium chloride (3%) is recommended.

  • Ingredients: Sodium phosphate, monobasic (NaH2PO4.xH2O), sodium phosphate, dibasic (Na2HPO4.xH2O), 0.5% sodium chloride (NaCl)

The pH of this buffers are 7.4 (0.1 M) and the osmolarity is 440 mosmols, so this buffers are hypertonic to most body fluids.

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Millonig's Phosphate Buffer, 0.2 M
  • Size:
    1 l
  • pH:
    7.4 (0.1 M)
  • Storage temperature:
    0...5 °C

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.