Orders LinkContactUsLinkComponent
Light Pipe Scintillators, Electron Microscopy Sciences
Cambridge Light Pipe Scintillator, Quartz, Straight, Up To 200 mm
Catalog # 100495-412
 :  82000-X
Light Pipe Scintillators, Electron Microscopy Sciences
Catalog # 100495-412
Light Pipe Scintillators, Electron Microscopy Sciences
Light Pipe Scintillators, Electron Microscopy Sciences
Cambridge Light Pipe Scintillator, Quartz, Straight, Up To 200 mm
Price
$1,650.57
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
  • Scintillator material coated directly on the light pipe which improves the signal
  • Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
 : Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
Specifications
  • Description:
    Cambridge Light Pipe Scintillator, Quartz, Straight, Up To 200 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)