Orders LinkContactUsLinkComponent
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Arsenic Grain
  100495-140
 :  80075-AS
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
  100495-140
 :  
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Arsenic Grain
 :  
$844.34
Each
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with a location map for standard identification. Grains of standard materials are available and they measure approximately 2 x 2 mm in size. They are supplied with fully authenticated certificates of analysis.
  • Description:
    Arsenic Grain
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)