Orders LinkContactUsLinkComponent
White Translucent Cutting Mat, Electron Microscopy Sciences
White Translucent Cutting Mat, Electron Microscopy Sciences
White Translucent Mat, 18 x 24"
Catalog # 100494-398
White Translucent Cutting Mat, Electron Microscopy Sciences
Catalog # 100494-398
Supplier:  
White Translucent Cutting Mat, Electron Microscopy Sciences
White Translucent Cutting Mat, Electron Microscopy Sciences
White Translucent Mat, 18 x 24"
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cutting Mats will protect all table-top surfaces from knife cuts. Its smooth surface permits straight, even cutting and the cut mark heals instantly. Each mat is printed with both 1/2" and 1" grid patterns to aid in measuring and aligning work. Available in translucent white, which allows for cutting and trimming on a light box surface.
Specifications
  • Description:
    White Translucent Mat, 18 x 24"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)