Order Entry
Canada
Orders LinkContactUsLinkComponent
Staining Dish Set for 30 Slides, Electron Microscopy Sciences

Staining Dish Set for 30 Slides, Electron Microscopy Sciences

Staining Dish Cover Only
Catalog # 100492-936
Supplier #:  70312-32
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Catalog # 100492-936
Supplier:  
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Staining Dish Set for 30 Slides, Electron Microscopy Sciences

Staining Dish Cover Only

Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These mix and match staining dish components offer greater flexibility in meeting your requirements. This staining dish accommodates 16, 20 and 30 slide racks.

  • Dish is made from soda-lime glass
  • Holds a variety of sizes: 3 x 1", 3 x 11/2 ", and 3 x 2"
  • Inside: 135 (H) x 102 (W) x 69 (D) mm
  • Outisde: 146 (H) x 113 (W) x 83 (D) mm

Delivery information: Set includes of one staining dish, one lid, one 30-slide stainless steel rack.

Specifications
  • Description:
    Staining Dish Cover Only

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.