Orders LinkContactUsLinkComponent
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Six counting unit tally counter
  100492-918
 :  70310-06
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
  100492-918
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Six counting unit tally counter
$1,279.61
Each
The desk tally counter comes with one, two, or six counting units.
One Counting Unit: A chrome plated with counter with a flange (21/2" dia.) for desktop mounting. The recording lever is on the right side and the reset knob on the top. The counter is ideal for serial sectioning work. Four digit place.
Two Counting Units: A Counter with two counting keys each with 0 to 999 range with a single reset knob it comes with removable tabs for top identification.
Six Counting Units: A counter with five keys and one totalize unit (0 to 999), which records the total number of strokes made on other keys. At 100 increments, totalizing unit rings bell. When totalizing unit records 100, each figure corresponding to individual keys automatically displays percentage. Two reset knobs, removable tabs on top of counter provide area for identification of clasification being counted.
  • Description:
    Six counting unit tally counter
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.