Orders LinkContactUsLinkComponent
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, 5DR, Yes
Catalog # 100489-530
Supplier #:  72646-05
Tooled Neck Vials, Electron Microscopy Sciences
Catalog # 100489-530
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, 5DR, Yes
Price
$518.33
Log in for availability
Each Pack of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
Specifications
  • Cap included:
    Yes
  • Description:
    Tooled Neck Vials, 5DR
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)