Large Glass Slide Storage Box, Electron Microscopy Sciences
Product Details & Documents
100-Slide Capacity:
These storage boxes are available in two sizes.
100-Slide Capacity:
Durable, wood frame, covered with heavy black embossed paper. The hinges and latches are Nickel plated. A numbered plastic insert provides spacing for the handling of the slides. The box accommodates 100 of 3x2" slides. The box measures 24.7 x 19.7 x 5.7 cm (9 3/4 x 7 4/5 x 2 1/4").
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
Specifications
- Description:100 Microscope Slide Box, 3x2" Slides
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)