Orders LinkContactUsLinkComponent
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish with Pyrex Dish
Catalog # 100501-410
Supplier #:  70540-50
Dissecting Dish, Electron Microscopy Sciences
Catalog # 100501-410
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish with Pyrex Dish
Price
$398.49
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A silicone pad placed inside a 150x20 mm deep plastic petri dish. The mat can be removed, washed, and returned to the dish for reuse. The pins come in 10 different coded colors.
Optional Pyrex Glass Petri dish is available for use when sterilization is needed.
Specifications
  • Description:
    Dissecting Dish with Pyrex Dish
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.