Syringe Filtration System, Electron Microscopy Sciences
Product Details & Documents
Syrings filters with high burst strength for assured performance. Manufactured to ISO 9002 and GMP standard and 100% integrity tested.
- For tissue culture and HPLC applications
- Membrane choices include:
- Regenerated cellulose (RC)
- Polyethersulfone (PES)
- Surfactant-free cellulose acetate (SFCA)
- Nylon (NY)
- 'PTFE' fluourocarbon resin
Syringe filter sizes include 4, 15, 25, or 26 mm diameter to handle volumes ranging from 0.5 to 100 ml. The 50 mm diameter 'PTFE' syringe filter are ideal for protecting the vacuum line and pump. These disposable filter units are ideal for tissue culture media, buffer solution, stain solution, biological fluids and most other aqueous solutions and reagents that are used daily in the EM lab.
Pore size and membrane type are printed on every syringe and the filters are certified non-cytotoxic and non-pyrogenic.
Packaging: Sterile syringe filters are individually blister-wrapped in cases of 50. Non sterile syringe filters are packaged in cases of 50. The 50 mm 'PTFE' syringe filter is supplied in a case of 12.
Specifications
- Ø:26 mm
- Housing material:Acrylic
- Pore size:0.45 µm
- Description:Syringe Filter SFCA
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.