Orders LinkContactUsLinkComponent
Micro Stirring Bars, Electron Microscopy Sciences
Micro Stirring Bars, Electron Microscopy Sciences
12.7 mm (½"), 3 mm (1/8 "), PTFE
Catalog # 100501-026
Supplier #:  64515-12
Micro Stirring Bars, Electron Microscopy Sciences
Catalog # 100501-026
Micro Stirring Bars, Electron Microscopy Sciences
Micro Stirring Bars, Electron Microscopy Sciences
12.7 mm (½"), 3 mm (1/8 "), PTFE
Price
$270.75
Log in for availability
Box of 12 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bars are coated with 'PTFE' white resin.
Used for stirring test tubes, micro-beakers, and more.
Specifications
  • Coating:
    PTFE
  • Ø:
    3 mm (1/8 ")
  • Length:
    12.7 mm (½")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)