Orders LinkContactUsLinkComponent
Embedding Rings, Electron Microscopy Sciences
Embedding ring, Yellow
  100500-980
 :  62350-Y
Embedding Rings, Electron Microscopy Sciences
  100500-980
Embedding Rings, Electron Microscopy Sciences
Embedding Rings, Electron Microscopy Sciences
Embedding ring, Yellow
$119.94
Box of 250 Each
Molded from the highest quality plastic, our Embedding Rings fit most base molds. For added convenience, there are four colors from which to choose Blue, Pink, Green, Yellow. The large writing surface further eases identification and handling. The base measures 1 5/8 x 1 1/8" and the ring measures 25 x 25 mm (1 x 1").
  • Color:
    Yellow
  • Description:
    Embedding ring
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.