Orders LinkContactUsLinkComponent
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipettes, Curved, 12 mL
  100500-758
 :  70986-20
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
  100500-758
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipettes, Curved, 12 mL
$195.51
Box of 50 Each
Transfer pipette that works as a syringe for liquid transfer. Each pipette has calibration markings from 0 to 12 ml in 0.2 ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
  • Inexpensive and disposable
  • Description:
    Transfer Pipettes, Curved, 12 mL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.