Orders LinkContactUsLinkComponent
Broken Crosslines, Electron Microscopy Sciences
Broken Crosslines, Electron Microscopy Sciences
NE56 Broken Crosslines Graticule, 16 mm
Catalog # 100499-026
Supplier #:  67985-16
Broken Crosslines, Electron Microscopy Sciences
Catalog # 100499-026
Broken Crosslines, Electron Microscopy Sciences
Broken Crosslines, Electron Microscopy Sciences
NE56 Broken Crosslines Graticule, 16 mm
Price
314,90 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Broken lines enable fine detail to be seen at the breaks. A thin boundary would be lost behind a continuous line.
Specifications
  • Description:
    NE56 Broken Crosslines Graticule
  • Size:
    16 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)