Orders LinkContactUsLinkComponent
Embedding Blocks, Electron Microscopy Sciences
Embedding Blocks, Electron Microscopy Sciences
Embedding Block 3, 38 x 38 x 19 mm
Catalog # 100498-058
Supplier #:  62363-03
Embedding Blocks, Electron Microscopy Sciences
Catalog # 100498-058
Embedding Blocks, Electron Microscopy Sciences
Embedding Blocks, Electron Microscopy Sciences
Embedding Block 3, 38 x 38 x 19 mm
Price
$202.05
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.
Specifications
  • Description:
    Embedding Block 3
  • Dimensions:
    38 x 38 x 19 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)