Orders LinkContactUsLinkComponent
Digimatic Calipers, Electron Microscopy Sciences
Digimatic Calipers, Electron Microscopy Sciences
Digimatic Caliper Absolute Function with Spc Output
Catalog # 100497-548
Supplier #:  62062-20
Digimatic Calipers, Electron Microscopy Sciences
Catalog # 100497-548
Digimatic Calipers, Electron Microscopy Sciences
Digimatic Calipers, Electron Microscopy Sciences
Digimatic Caliper Absolute Function with Spc Output
Price
$931.70
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All metal head with a plastic housing.
Provides a digital readout to 0.0005" / 0.01 mm for error-free reading and can measure steps. Function: zero setting, data hold, low battery alarm and data output. Provided with a data saving output port for connection with the SPC system. Powered by a single SR-44 battery. Battery life: approximately two years under normal use.
Specifications: Range: 0 – 6" / 0 - 150 mm; LCD resolution: 0.0005" / 0.0; Accuracy: ±0.001
Specifications
  • Description:
    Digimatic Caliper Absolute Function with Spc Output
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.