Orders LinkContactUsLinkComponent
BEEM™ Block Storage, Electron Microscopy Sciences
BEEM™ Block Storage, Electron Microscopy Sciences
Block Storage
Catalog # 100492-870
Supplier #:  69922-01
BEEM™ Block Storage, Electron Microscopy Sciences
Catalog # 100492-870
BEEM™ Block Storage, Electron Microscopy Sciences
BEEM™ Block Storage, Electron Microscopy Sciences
Block Storage
Price
$19.86
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A two tone color coded plastic box with insert.
  • Allows storage of up to 10 blocks in the numbered cavities
LxWxH: 57x45x12.5 mm (2 1/3 x 1 3/4 x 1/2")
Specifications
  • Description:
    Block Storage
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)