Pocket Measuring Microscopes, Electron Microscopy Sciences
As small as a fountain pen, the pocket measuring microscope has a standard scale is incorporated in its lens barrel, giving an effecient brightness and excellent definition. It can be focused in the same way as a conventional microscope.
- 25x magnification has 3.30 mm field of view, 3 mm measuring range and minimal scale division of 0.005
- 50x magnification has 2.00 mm field of view, 1.6 mm measuring range and minimal scale division of 0.02
Dimensions: 12.40 x 127 mm
- Magnification:50×
- Description:Pocket Measuring Microscope
- Cat. No.:100491-450
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.