Orders LinkContactUsLinkComponent
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grid slides, round well
Catalog # 100490-308
Supplier #:  63408-01
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Catalog # 100490-308
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grid slides, round well
Price
$326.23
Log in for availability
Each Pack of 72
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These 3×1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents.
  • Each square grid fills one low-powered-field (approximately 100× magnification)
  • Easy to locate an organism or cell every time
  • Eighty one squares
The slides are available with coatings of Poly-L-Lysine or Silane, or they can be purchased plain (uncoated). The grid measures 2 mm square, 16 mm total. Numbers 1-8 and alpha A-H.
Specifications
  • Description:
    Grid slides, round well
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)