Orders LinkContactUsLinkComponent
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, 3DR, Yes
  100489-526
Supplier #:  72646-03
Tooled Neck Vials, Electron Microscopy Sciences
  100489-526
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, 3DR, Yes
$327.13
Each Pack of 144
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
  • Cap included:
    Yes
  • Description:
    Tooled Neck Vials, 3DR
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)