Orders LinkContactUsLinkComponent
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
Embedding Capsule Bottle Neck Tip
Catalog # 100501-200
Supplier #:  69912-01
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
Catalog # 100501-200
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
Embedding Capsule Bottle Neck Tip
Price
$23.88
Log in for availability
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Available in two specially shaped capsules which are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. The bottle neck capsule produces a 7.9 mm OD block with a hemihyperpoloid (rather than faced) tip. The conical capsule produces a 7.9 mm OD block with a conical (rather than faced) tip.
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Specifications
  • Description:
    Embedding Capsule Bottle Neck Tip
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.