Orders LinkContactUsLinkComponent
Capsule Molds, Electron Microscopy Sciences
Capsule Molds, Electron Microscopy Sciences
Capsule Mold Type A, 10 Cavities
Catalog # 100491-596
Supplier #:  70150
Capsule Molds, Electron Microscopy Sciences
Catalog # 100491-596
Capsule Molds, Electron Microscopy Sciences
Capsule Molds, Electron Microscopy Sciences
Capsule Mold Type A, 10 Cavities
Price
$49.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. They are all reusable and produce embedding blocks of numerous shapes and sizes with a wide variety of tips. We also offer a mold which produces blocks to be used for re-embedding.
Specifications
  • Description:
    Capsule Mold Type A, 10 Cavities
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.