Orders LinkContactUsLinkComponent
Titramax Vibrating Microtiterplate Shaker^ Model 100. Accommodates up to 4 microtiterplates. Analog. Speed range: 150-1350rpm. Maintenance-free motor. Self-resets to protect against overheating. Orbit size: 1.5mm. Max. load capacity: 2kg. 24.5Wx31Dx12.5Hcm. Shipping weight: 5.5kg. 115V, 60Hz, 15W.
Titramax Vibrating Microtiterplate Shaker^ Model 100. Accommodates up to 4 microtiterplates. Analog. Speed range: 150-1350rpm. Maintenance-free motor. Self-resets to protect against overheating. Orbit size: 1.5mm. Max. load capacity: 2kg. 24.5Wx31Dx12.5Hcm. Shipping weight: 5.5kg. 115V, 60Hz, 15W.
Catalog # CA11009-540
Titramax Vibrating Microtiterplate Shaker^ Model 100. Accommodates up to 4 microtiterplates. Analog. Speed range: 150-1350rpm. Maintenance-free motor. Self-resets to protect against overheating. Orbit size: 1.5mm. Max. load capacity: 2kg. 24.5Wx31Dx12.5Hcm. Shipping weight: 5.5kg. 115V, 60Hz, 15W.
Catalog # CA11009-540
Supplier:  
Titramax Vibrating Microtiterplate Shaker^ Model 100. Accommodates up to 4 microtiterplates. Analog. Speed range: 150-1350rpm. Maintenance-free motor. Self-resets to protect against overheating. Orbit size: 1.5mm. Max. load capacity: 2kg. 24.5Wx31Dx12.5Hcm. Shipping weight: 5.5kg. 115V, 60Hz, 15W.
Titramax Vibrating Microtiterplate Shaker^ Model 100. Accommodates up to 4 microtiterplates. Analog. Speed range: 150-1350rpm. Maintenance-free motor. Self-resets to protect against overheating. Orbit size: 1.5mm. Max. load capacity: 2kg. 24.5Wx31Dx12.5Hcm. Shipping weight: 5.5kg. 115V, 60Hz, 15W.
Discontinued
Product Details & Documents