Round Bottom Flasks
10 ml, 10/19, Round
Catalog # CA11009-132
Supplier: Heidolph NA, LLC
Supplier #: 015890690CA
Jump to:
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
- [ST] joint no.:10/19
- Capacity:10 ml
- Bottom style:Round