Orders LinkContactUsLinkComponent
Round Bottom Flasks
Round Bottom Flasks
50 ml, 24/29, Round
Catalog # CA11009-122
Supplier:   Heidolph NA, LLC
Supplier #:  015890640CA
Round Bottom Flasks
Catalog # CA11009-122
Supplier:  Heidolph NA, LLC
Round Bottom Flasks
50 ml, 24/29, Round
Price
$656.26
Log in for availability
Pack of 5
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
  • [ST] joint no.:
    24/29
  • Capacity:
    50 ml
  • Bottom style:
    Round