Round Bottom Flasks
250 ml, 19/26, Round
Catalog # CA11009-112
Supplier: Heidolph NA, LLC
Supplier #: 015890590CA
Jump to:
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
- [ST] joint no.:19/26
- Capacity:250 ml
- Bottom style:Round