Prefilled Bouin's Solution, Azer Scientific
Prefilled Bouin's Solution, Half Filled, 20 ml
Supplier: VWR International
Prefilled Bouin's Solution, Azer Scientific
Catalog # 89217-510
Supplier: VWR International
CAS Number:
Jump to:
Product Details & Documents
Bouin's fixative is a saturated picric acid-formalin solution that is useful for preserving and fixing delicate tissues and gastrointestinal tract specimens.
- Available in 20, 40 and 60 ml containers each half-filled
- 95 kPa compliant specimen containers
Specifications
Specifications
- Description:Prefilled Bouin's Solution, Half Filled
- Size:20 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



