Bedside Carts, Lakeside®
Bedside 3-Drawer
Catalog # 89210-186
Supplier: Lakeside
Supplier #: BV06
Jump to:
Product Details & Documents
Value-driven and economically designed carts are perfect for the cost-conscious, budget-driven healthcare facility
Color selection is limited to beige only. Push handle for increased control. Rubber corner bumpers extend out to protect walls and furniture. Epoxy powder coating resists marring, chipping and peeling. Roller bearing drawer slides prevent drawer sticking.
Specifications
- Description:Bedside 3-Drawer