Order Entry
Canada
ContactUsLinkComponent
Extracta Dna Prep 25Ml
Extracta™ DNA Prep for PCR, Quantabio
Catalog # CA97065-350
Supplier:  Quantabio
CAS Number:  
undefined
Extracta™ DNA Prep for PCR, Quantabio
Catalog # CA97065-350
Supplier:  Quantabio
Supplier Number:  95091-025
CAS Number:  

Specifications

  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Extracta™ DNA prep for PCR
  • Size:
    25 ml
  • Description:
    Extracta™ DNA prep for PCR
  • Cat. no.:
    CA97065-350
  • Supplier No.:
    95091-025

Specifications

About this item

Extracta DNA Prep for PCR is a two-component reagent kit for rapid extraction of PCR-ready genomic DNA from a variety of tissues. Samples are processed in less than 30 minutes with minimal hands-on time and technical skill.

  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, High Resolution Melt Analysis, endpoint PCR and other applications
  • Works with wide range of tissue including hair, buccal swabs and mouse tails
  • Optional 10 minute protocol for high-throughput applications

Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.

The Extracta™ DNA prep for PCR consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva. The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.

Ordering information: DNA Prep kit includes extraction reagent and stabilization buffer.
Frequently Bought Together