Orders LinkContactUsLinkComponent
pH Electrode Solutions, WTW
Pepsin Cleaning Solution, 3 x 250 ml, PEP/pH
  89207-612
 :   WTW
 :  109648
pH Electrode Solutions, WTW
  89207-612
 :  WTW
 :  
pH Electrode Solutions, WTW
Pepsin Cleaning Solution, 3 x 250 ml, PEP/pH
 :  
$352.17
Each (250ml)
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
  • Description:
    Pepsin Cleaning Solution, 3 x 250 ml, PEP/pH