Order Entry
Canada
Orders LinkContactUsLinkComponent
Germicidal Bulbs

Germicidal Bulbs

Bulb, Linear Double Ended, Light Sources, Rapid Start (Circuitry), T5, 39 W, 253.7 nm, FA8/slimline, single pin, 7500 h , 843.28 mm, 15.875 mm
Catalog # 47736-183
Germicidal Bulbs
Catalog # 47736-183
Supplier:  
Germicidal Bulbs
Germicidal Bulbs

Bulb, Linear Double Ended, Light Sources, Rapid Start (Circuitry), T5, 39 W, 253.7 nm, FA8/slimline, single pin, 7500 h , 843.28 mm, 15.875 mm

Discontinued
Product Details & Documents

Various germicidal bulbs.

Specifications
  • Ø:
    15.875 mm
  • Description:
    Bulb, Linear Double Ended, Light Sources, Rapid Start (Circuitry)
  • Length:
    843.28 mm
  • Bulb life:
    7500 h
  • Base:
    FA8/slimline, single pin
  • Model:
    T5
  • Power:
    39 W
  • Wavelength:
    253.7 nm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.