NxGen™ M-MulV Reverse Transcriptase, Biosearch Technologies
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Lucigen’s NxGen™ M-MulV Reverse Transcriptase is outstanding in cDNA synthesis.
M-MuLV Reverse Transcriptase is an RNA-dependent DNA polymerase which shows no measurable 3′→5′ proofreading function. This enzyme can copy a single-stranded DNA template or perform cDNA synthesis by extending a DNA primer annealed to an RNA template.
Performance Specifications: 1) Unit Concentration: 200,000 U/mL. 2) Purity (SDS-PAGE): >99%. 3) SS Exonuclease: 2,000 U <5.0% released. 4) DS Exonuclease: 2,000 U <0.5% released. 5) Endonuclease: 2,000 U <10% converted. 5) E. coli 16S rDNA Contamination: 2,000 U <10 copies. 6) Storage: -20°C.
Source: A recombinant E. coli strain carrying the Moloney-Murine Leukemia Virus Reverse Transcriptase gene.
Ordering information: Supplied at 200,000 U/mL in 50 mM Tris-HCl, 150 mM NaCl, 0.1 mM EDTA, 1 mM dithiothreitol, 0.1% NP-40 Alternative, 50% glycerol, pH 7.6 @ 25°C. Also provided is 10X M-MuLV RT Buffer which in 1X form is composed of 50 mM Tris-HCl, 75 mM KCl, 3 mM MgCl2, 10 mM dithiothreitol, pH 8.3 @ 25°C. 250,000 U is supplied as 5 × 50,000 U. Unit definition: 1 unit is defined as the amount of enzyme required to incorporate 1 nmol of dTTP into acid insoluble material in 10 minutes at 37°C using poly r(A)/oligo (dT) as a substrate.
Specifications
Specifications
- Description:NxGen™ M-MulV Reverse Transcriptase
- Units:50000
- No. of reactions:50000
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



