Screw Caps, MicroSolv™
PP, Bonded Silicone/PTFE, pre-slit, 9-425 mm, Blue, Screw cap, knurled
Catalog # 97051-426
Supplier: MicroSolv Technology Corp
Supplier #: 9502S-30C-B
Jump to:
Product Details & Documents
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Documents
Specifications
- Cap Size:9-425 mm
- Cap type:Screw cap, knurled
- Septum:Bonded Silicone/PTFE, pre-slit
- Material:PP
- Cap color:Blue