Orders LinkContactUsLinkComponent
ELGA Deionization Systems
ELGA Deionization Systems
Deionizer replacement cartridge, for Vision 125, filter
Catalog # CA89204-278
Supplier:   ELGA CA
Supplier #:  VC125
ELGA Deionization Systems
Catalog # CA89204-278
Supplier:  ELGA CA
ELGA Deionization Systems
ELGA Deionization Systems
Deionizer replacement cartridge, for Vision 125, filter
Price
$373.65
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
ELGA’s range of wall mounted deionization systems produce from 30 to 90 liters per hour of high purity water for a wide range of laboratory and general manufacturing applications.
  • Cost-effective
  • Easy monitoring
  • Built-in meters
  • Water Quality: Type II
The deionizers are cost-effective stand-alone systems that provide low to medium volumes of primary or deionized grade water. Water quality is easy to monitor, with built in meters or colour change indicators.
Specifications
  • Description:
    Deionizer replacement cartridge, for Vision 125, filter