VWR® Refrigerated Circulating Baths
Product Details & Documents
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40° to 200°C (depending on the controller).
- Offer precise temperature control
- Available in a wide range of sizes
- Three controller options
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for many applications.
The large, LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Certifications: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
Ordering information: Refrigerated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8, 1/4" and 3/16" (.95, .63 and .47 cm) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.
Services: Every VWR® ISO 17025 NIST Traceable Pre-calibrated Circulating Bath is calibrated and tested by an ISO 17025 accredited lab to ensure the highest standard of accuracy and traceability. Two options are available: Option 1: Standard Pre-Calibrated, Model circulating bath is calibrated and tested at both 37°C and 70°C. Option 2: Customer specified pre-calibrated circulating bath provides a choice of 3 different temperature points based on your research requirements. Simply order the stated catalog number and you will receive a protective box containing a work order form allowing you to specify your own calibration temperature points. After completing the work order, you can mail the box using a pre-paid return shipping label or you can scan and email the information to a specified E-mail address. The Avantor calibration team will calibrate the circulating bath based on your instruction.
VWRCATALYST™ can provide an array of services, such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.
Documents
89202-962 Manual_English
Manual
Circulating Bath
Videos
Specifications
- Maximum Flow Rate:5.3 gpm (20.1 l/min.)
- Temperature range:–20...+200 °C
- Description:7L Space-Saving, Refrigerated Circulator with Advanced Digital Controller, -20°C
Complementary Products
Product Information
- Catalog #DescriptionProbeRangeAvailabilityPrice
- 89369-150Platinum high-accuracy freezer themometerStainless-steel probe with handle−100.00...+199.99 °C (−148.00...+391.99 °F)Log in for availabilityEach - $574.74
Accessories
Product Information
- Catalog #SizeFor Use WithAvailabilityPrice
- PolyScience polytherm S250PolyScience polytherm S25013270-7923.8 l (1 gal.)VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsLog in for availabilityEach (1US Gal) - $3,876.06
Product Information
- Catalog #SizeFor Use WithAvailabilityPrice
- PolyScience polytherm S150PolyScience polytherm S15013270-7943.8 l (1 gal.)VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsLog in for availabilityEach (1US Gal) - $2,893.65
Product Information
- Catalog #SizeFor Use WithAvailabilityPrice
- PolyScience polycool HC -50, Freezing Point –56 °CPolyScience polycool HC -50, Freezing Point –56 °C13272-0343.8 l (1 gal.)VWR Refrigerated Circulating BathsLog in for availabilityEach (1US Gal) - $424.81
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- PolyScience polyclean CLARIFIERPolyScience polyclean CLARIFIER71002-500VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems8 oz bottle of polyclean CLARIFIER; concentrated; treats approximately 200 gallons (757 liters)Log in for availabilityEach (8oz) - $83.31
Product Information
- Catalog #Temperature RangeAvailabilityPrice
- Ethylene glycol, 3.8 l (1 gal.)Ethylene glycol, 3.8 l (1 gal.)71002-504–25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled waterLog in for availabilityEach (1US Gal) - $253.34
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- PolyScience polyclean CLARIFIERPolyScience polyclean CLARIFIER71002-502VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systemscase of 12 (8 oz bottles); polyclean CLARIFIER; concentrated; treats appromixately 200 gallons (757 liters)Log in for availabilityCase of 12 (1/Itm/Bx) - $952.76
Product Information
- Catalog #Temperature RangeAvailabilityPrice
- Propylene glycol, 3.8 l (1 gal.)Propylene glycol, 3.8 l (1 gal.)71002-506–20...+100 °C (–4...+212 °F), when mixed 50/50 with distilled waterLog in for availabilityEach (1US Gal) - $457.69
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 6" External Pt100 Probe with 7.6 m (25') length cable6" External Pt100 Probe with 7.6 m (25') length cable89201-006VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationLog in for availabilityEach - $1,029.25
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 6" External Pt100 Probe with 3 m (10') length cable6" External Pt100 Probe with 3 m (10') length cable89201-004VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationLog in for availabilityEach - $743.10
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 6" External Pt100 Probe with 15 m (50') length cable6" External Pt100 Probe with 15 m (50') length cable89201-008VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationLog in for availabilityEach - $1,552.33
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- RS232 CableRS232 Cable89201-010VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Log in for availabilityEach - $101.37
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 1/4" ID Buna N Tubing1/4" ID Buna N Tubing89201-056VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems–40...120 °C, Order in Continuous Lengths Of 3' (1 m)Log in for availabilityEach - $33.01
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- Ethernet CableEthernet Cable89201-012VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Log in for availabilityEach - $88.59
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- Insulation for 1/4" TubingInsulation for 1/4" Tubing89201-062VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsOrder in Continuous Lengths of 2 Yards (6 ft.)Log in for availabilityEach - $143.30
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- Tube ClampsTube Clamps89201-076VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsFor 1/4 and 3/8" ID Tubing, Stainless SteelLog in for availabilityEach - $16.46