Orders LinkContactUsLinkComponent
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining Jars, Slide Staining Jars: 64L x 76W x 92H mm (21/2 x 3 x 35/8"), Slide Staining Rack: 60L x 64W x 97H mm (23/8 x 21/2 x 313/16"), Blue
Catalog # 87000-120
Supplier #:  M900-12B
EasyDip™ Slide Staining System, Simport Scientific
Catalog # 87000-120
Supplier:  
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining Jars, Slide Staining Jars: 64L x 76W x 92H mm (21/2 x 3 x 35/8"), Slide Staining Rack: 60L x 64W x 97H mm (23/8 x 21/2 x 313/16"), Blue
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
JAR SLIDE-STAINING EASY-DIP BLUE CS2 Catalog #102097-484
Product 102097-484 is a replacement for 87000-120 and has the same packaging and specifications, but may be a different quantity.
EASYDIP SLIDE STAIN JAR BLUE Catalog #CA87000-120
Product CA87000-120 is a replacement for 87000-120 and has the same packaging and specifications, but may be a different quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The slide staining system is made of acetal plastic.
  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide stable base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications
The staining jar is made of resistant acetal plastic, and will not break like most glass jars do. It will resist most staining agents including alcohol and xylene (but not phenol, iodine, or ferric chloride). The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80 ml. Easy to clean and no metals to corrode. Ideal for special stains, frozen sections, and special processes.
The system is not autoclavable. Volume is 80 ml. System dimensions are 64×76×92 mm (2.5×3×3.625").
Ordering information: The staining jars and staining rack are sold separately. The EasyDip™ Kit with Rack includes one anodized aluminum holder along with six assorted color jars (two white ones) and one M905-12DGY Slide Staining Rack. The EasyDip™ Aluminum Holder is designed to hold six EasyDip™ slide staining jars. The assorted kit includes one jar of each color.
Specifications
  • Color:
    Blue
  • Description:
    EasyDip™ Slide Staining Jars
  • Dimensions:
    Slide Staining Jars: 64L x 76W x 92H mm (21/2 x 3 x 35/8"), Slide Staining Rack: 60L x 64W x 97H mm (23/8 x 21/2 x 313/16")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.