Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACETONITRILE LC/MS REAGENT 4L
Catalog # TXCAJT9829-3
Supplier:
AVANTOR PERFORMANCE MATERIAL LLC
MOC:
Non-Regulated
Certificates
Expand image
ACETONITRILE LC/MS REAGENT 4L
Catalog # TXCAJT9829-3
Supplier:
AVANTOR PERFORMANCE MATERIAL LLC
Supplier Number:
BAKR9829-03
MOC:
Non-Regulated
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Additional Product Details
9829 Product Specifications
Documentation
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH