Orders LinkContactUsLinkComponent
Casters for Modular Workstations
Casters for Modular Workstations
Low profile casters lower worksurfaces, 200 lbs.
Catalog # 89202-112
Casters for Modular Workstations
Catalog # 89202-112
Supplier:  
Casters for Modular Workstations
Casters for Modular Workstations
Low profile casters lower worksurfaces, 200 lbs.
Discontinued
Product Details & Documents
Make any Sovella lab bench a mobile lab station by adding casters! Work is not always confined to one location.
  • Contain foot activated total lock brake
  • Easy swiveling and smooth rolling
  • Durable zinc plated finish
Ordering information: Casters sold in sets of four. Installation is required. Assembly videos available.
Specifications
  • Description:
    Low profile casters lower worksurfaces
  • Load capacity:
    200 lbs.