Orders LinkContactUsLinkComponent
TOC Cleaning Validation Kits, Texwipe® | Avantor
TOC Cleaning Validation Kits, Texwipe®
Kit, incl. 12 × 40 ml clear vials bonded septa caps (certified <10 µg/L TOC), 24 × TX714K Low TOC Alpha® Sampling Swabs (certified <50 µg/L TOC), and 12 blank vial labels
  TWTX3340
 :   Texwipe
Fournisseur #:  TX3340
TOC Cleaning Validation Kits, Texwipe®
  TWTX3340
 :  Texwipe
TOC Cleaning Validation Kits, Texwipe®
TOC Cleaning Validation Kits, Texwipe®
Kit, incl. 12 × 40 ml clear vials bonded septa caps (certified <10 µg/L TOC), 24 × TX714K Low TOC Alpha® Sampling Swabs (certified <50 µg/L TOC), and 12 blank vial labels
$281.16
Each Case of 18
Texwipe’s Cleaning Validation Kits were designed to simplify sampling as part of a cleaning validation protocol. The swabs and vials are further cleaned using a proprietary process to make these kits excellent for use in Total Organic Carbon (TOC) analysis.
  • Swab head is cleanroom laundered, double-layered polyester knit
  • Break-away handle allows the swab head to be placed into a sampling vial with minimal handling
Kits contain vials, swabs, and labels to sample multiple places in the production area. Prewashed TOC vials have been manufactured, analyzed, and certified to virtually elimnate all background TOC. Swab has a large head that provides firm support for sampling broad areas and flat surfaces. Can be used with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, Ol Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Packaging: Kits are packaged in cleanroom-compatible polypropylene boxes for efficient transport of samples from production to laboratory with minimal chance of contamination.
Documents
  • Description:
    Kit, incl. 12 × 40 ml clear vials bonded septa caps (certified <10 µg/L TOC), 24 × TX714K Low TOC Alpha® Sampling Swabs (certified <50 µg/L TOC), and 12 blank vial labels
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.