Orders LinkContactUsLinkComponent
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
25 ml, Pear-shaped flask, threaded neck, O.D×H: 80×39 mm
Catalog # KT747540-2520
 :  747540-2520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
Catalog # KT747540-2520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
25 ml, Pear-shaped flask, threaded neck, O.D×H: 80×39 mm
Price
$45.41
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
These flasks have short necks for easy retrieval of residues or reaction products.
  • Accessory for Microscale MICROFLEX® threaded kits
  • Thread size: 20-400
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Specifications
  • Capacity:
    25 ml
  • Description:
    Pear-shaped flask, threaded neck, O.D×H: 80×39 mm