Order Entry
Canada
Orders LinkContactUsLinkComponent
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences

KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences

5 ml, Pear-shaped flask, threaded neck, O.D×H: 25×50 mm
Catalog # KT747540-0520
Supplier #:  747540-0520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
Catalog # KT747540-0520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences

5 ml, Pear-shaped flask, threaded neck, O.D×H: 25×50 mm

Price
$32.58
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

These flasks have short necks for easy retrieval of residues or reaction products.

  • Accessory for Microscale MICROFLEX® threaded kits
  • Thread size: 20-400

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Specifications
  • Capacity:
    5 ml
  • Description:
    Pear-shaped flask, threaded neck, O.D×H: 25×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.