KIMBLE® TLC Cylindrical Developing Tanks, DWK Life Sciences
Tank with PE Lid, 2.54×7.62 cm plates
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 416170-1118
KIMBLE® TLC Cylindrical Developing Tanks, DWK Life Sciences
Catalog # KT416170-1118
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Cylindrical glass tanks for the development of 1x3" to 10x20cm plates
These tanks provide maximum vapor phase saturation with a minimum amount of solvent.
Specifications
- Description:Tank with PE Lid
- For use with:2.54×7.62 cm plates
Accessories
Product Information
- Catalog #AvailabilityPrice
- Polyethylene cap, for 5×10 and 5×20 cm tanksPolyethylene cap, for 5×10 and 5×20 cm tanksKT416170-1125Log in for availabilityPack of 12 - 163,80 $
Product Information
- Catalog #AvailabilityPrice
- Polyethylene cap, for 2.54×7.62 cm tanksPolyethylene cap, for 2.54×7.62 cm tanksKT416170-1124Log in for availabilityPack of 12 - 163,80 $
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



