Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
WIPER 100% IPA 9X9 BG50 CS12
Catalog # CATWTX1082
Supplier:
ITW Texwipe
SDS
Certificates
Expand image
WIPER 100% IPA 9X9 BG50 CS12
Catalog # CATWTX1082
Supplier:
ITW Texwipe
Supplier Number:
TX1082
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_TX1082
TXTWTX1082_193904
TXTWTX1082_194004
TXTWTX1082_194404
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning