Order Entry
Canada
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded

J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded

± 0.04, 25 °C, Cubetainer®, 4 L, 7.00, Yellow
Catalog # CAJT5656-3
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
Catalog # CAJT5656-3
Supplier:  
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded

± 0.04, 25 °C, Cubetainer®, 4 L, 7.00, Yellow

CAS Number:  
MOC:  Non-Regulated, With Information
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER COLOR CODED PH7 4-L Catalog #JT5656-3

Product JT5656-3 is a replacement for CAJT5656-3 and has the same packaging and specifications, but may be a different quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This BAKER ANALYZED® Reagent is a phosphate buffer solution for laboratory, research, and/or manufacturing uses.

  • Contains no presevative

Each lot is compared to NIST Standard Reference Buffer.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Cubetainer®
  • Color:
    Yellow
  • Size:
    4 L
  • Reference temperature:
    25 °C
  • pH:
    7.00
  • pH accuracy:
    ± 0.04

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.