Order Entry
Canada
Orders LinkContactUsLinkComponent
J.T.Baker®, pH Buffer Solutions

J.T.Baker®, pH Buffer Solutions

Cubetainer®, 4 L, 10.00, Blue
Catalog # CAJT5655-3
Supplier #:  BAKR5655-03
J.T.Baker®, pH Buffer Solutions
Catalog # CAJT5655-3
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
J.T.Baker®, pH Buffer Solutions
J.T.Baker®, pH Buffer Solutions

Cubetainer®, 4 L, 10.00, Blue

CAS Number:  
MOC:  Non-Regulated, With Information
Price
$353.09
Log in for availability
Each (4l) Case of 4 (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

High quality buffer solutions ensure accurate pH measurements time after time.

  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.

Solutions are available in both clear and color-coded varieties.

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Certifications: Certified traceable to NIST Standard Reference Buffers.

Specifications

Specifications

  • Pack type:
    Cubetainer®
  • Color:
    Blue
  • Size:
    4 L
  • pH:
    10.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.