J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Phosphate
Product Details & Documents
This BAKER ANALYZED® Reagent is a phosphate buffer solution for laboratory, research, and/or manufacturing uses.
- Contains no antimicrobial preservative
Each lot is compared to NIST Standard Reference Buffer.
Documents
Certificates
Search by Lot Number5608 Product Specifications
Additional Product Details
spec_JT5608-3
Product specification
Specifications
Specifications
- Pack type:Cubetainer®
- Color:Colorless
- Size:4000 ml
- Reference temperature:25 °C
- pH:7.00
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



